Benzyl isothiocyanate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
252492-25G
252492-5G
SIAL252492-25GEA
2600
CZK
SIAL252492-25G
SIAL252492-5G
Benzyl isothiocyanate, Sigma-Aldrich®
Benzyl isothiocyanate
Vzorec:
C₈H₇NS Bod varu: 243 °C (1013 hPa) Bod tání: 24 °C Hustota: 1,084 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004819 CAS číslo: 622-78-6 EINECS: 210-753-9 UN: 2206 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...