Tisk…

p-Tolyl isocyanate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Methylphenyl isocyanate, 1-Isocyanato-4-methylbenzene

M-Clarity™ quality level = MQ100

143634-100G 143634-25G 143634-5G
SIAL143634-100GEA 10250 CZK
SIAL143634-100G SIAL143634-25G SIAL143634-5G
p-Tolyl isocyanate, Sigma-Aldrich®
p-Tolyl isocyanate
Vzorec: C₈H₇NO
MW: 133,15 g/mol
Bod varu: 193…195 °C (1013 hPa)
Hustota: 1,056 g/cm³ (20 °C)
MDL Number: MFCD00002029
CAS číslo: 622-58-2
EINECS: 210-743-4
UN: 2206
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR