m-Tolyl isocyanate ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 99%
CAS No.: 621-29-4
Molecular Formula: C8H7NO
Formula Weight: 133.15
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.5305
Odour: Pungent
Boiling Point: 75-76°/12mm
Beilstein Reference: 386149
UN No.: 2206
MDL No.: MFCD00002021
Vzorec:
C₈H₇NO Bod varu: 75…76 °C (12 mmHg) Bod tání: 195…198 °C Hustota: 1,033 Bod vzplanutí: 65 °C (149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002021 CAS číslo: 621-29-4 EINECS: 210-676-0 UN: 2206 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...