Benzyl(dimethyl)amine for synthesis, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA
M-Clarity™ quality level = MQ200
8.03206.0250
8.03206.1000
8.03206.0250EA
1090
CZK
8.03206.0250
8.03206.1000
Benzyl(dimethyl)amine for synthesis, Sigma-Aldrich®
Benzyl(dimethyl)amine
Vzorec:
C₆H₅CH₂N(CH₃)₂ MW: 135,21 g/mol Bod varu: 183 °C (1013 hPa) Bod tání: –75 °C Hustota: 0,915 g/cm³ (20 °C) Bod vzplanutí: 53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008329 CAS číslo: 103-83-3 EINECS: 203-149-1 UN: 2619 ADR: 8 (3),II |
Výsledky specifikačního testu
Solubility | 8 g/l (20 °C) |
Melting Point | -75 °C |
Molar Mass | 135.21 g/mol |
Boiling Point | 180 - 183 °C (1013 hPa) |
Vapor Pressure | 2.4 hPa (20 °C) |
Flash Point | 55 °C |
Refractive Index | 1.5011 (20 °C, 589 nm) |
Explosion Limit | 0.9 - 6.3 %(V) |
Density | 0.90 g/cm3 (20 °C) |
pH | 10 (10 g/l, H2O, 20 °C) (saturated solution) |
Ignition Point | 410 °C |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...