Benzyl(dimethyl)amine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98+%
CAS No.: 103-83-3
Molecular Formula: C9H13N
Formula Weight: 135.21
Flash Point: 54°(129°F)
Refractive Index: 1.5010
Melting Point: -75°
Boiling Point: 183-184°
Beilstein Reference: 1099620
UN No.: 2619
MDL No.: MFCD00008329
Vzorec:
C₆H₅CH₂N(CH₃)₂ Bod varu: 183…184 °C Bod tání: –75 °C Hustota: 0,900 Bod vzplanutí: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008329 CAS číslo: 103-83-3 EINECS: 203-149-1 UN: 2619 ADR: 8,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...