2-Iodobenzyl alcohol ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
L15126.06
L15126.14
L15126.06EA
1240
CZK
L15126.06
L15126.14
2-Iodobenzyl alcohol ≥99%
2-Iodobenzyl alcohol
Purity: 99%
CAS No.: 5159-41-1
Molecular Formula: C7H7IO
Formula Weight: 234.04
Melting Point: 89-92°
Beilstein Reference: 2079487
Vzorec:
C₇H₇IO MW: 234,04 g/mol Bod varu: 145 °C (10 mmHg) Bod tání: 89…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004610 CAS číslo: 5159-41-1 EINECS: 225-933-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...