Tisk…

4-Iodo-2-nitrotoluene ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

A18607.06 A18607.14
A18607.06EA 2910 CZK
A18607.06 A18607.14
4-Iodo-2-nitrotoluene ≥97%
4-Iodo-2-nitrotoluene

Purity: 97% CAS No.: 41252-97-5 Molecular Formula: C7H6INO2 Formula Weight: 263.03 Melting Point: 58-62° MDL No.: MFCD00051090

Vzorec: C₇H₆INO₂
Bod tání: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00051090
CAS číslo: 41252-97-5
EINECS: 255-279-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR