Tisk…

2-Iodo-m-toluic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

560499-1G 560499-5G
SIAL560499-1GEA 1820 CZK
SIAL560499-1G SIAL560499-5G
2-Iodo-m-toluic acid, Sigma-Aldrich®
2-Iodo-m-toluic acid
Vzorec: C₈H₇IO₂
MW: 262,05 g/mol
MDL Number: MFCD00079764
CAS číslo: 108078-14-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR