(4-Iodophenyl)acetonitrile ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-(4-Iodophenyl)acetonitrile, 4-Iodobenzyl cyanide, 4-Iodophenylacetonitrile
L15119.03
L15119.06
L15119.03EA
2250
CZK
L15119.03
L15119.06
(4-Iodophenyl)acetonitrile ≥97%
(4-Iodophenyl)acetonitrile
Purity: 97%
CAS No.: 51628-12-7
Molecular Formula: C8H6IN
Formula Weight: 243.05
Flash Point:
Vzorec:
C₈H₆IN MW: 243,05 g/mol Bod varu: 300 °C (1013 hPa) Bod tání: 52…54 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00060306 CAS číslo: 51628-12-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...