Tisk…

(3-Iodophenyl)acetonitrile, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Iodobenzyl cyanide, 2-(3-Iodophenyl)acetonitrile

M-Clarity™ quality level = MQ100

646415-1G 646415-5G
SIAL646415-1GEA 2170 CZK
SIAL646415-1G SIAL646415-5G
(3-Iodophenyl)acetonitrile, Sigma-Aldrich®
(3-Iodophenyl)acetonitrile
Vzorec: C₈H₆IN
MW: 243,05 g/mol
Bod varu: 138 °C (12 mmHg)
Bod tání: 33…35 °C
MDL Number: MFCD00040890
CAS číslo: 130723-54-5
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR