(3-Iodophenyl)acetonitrile ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-Iodobenzyl cyanide, 2-(3-Iodophenyl)acetonitrile
L15118.03
L15118.06
L15118.03EA
2300
CZK
L15118.03
L15118.06
(3-Iodophenyl)acetonitrile ≥97%
(3-Iodophenyl)acetonitrile
Purity: 97%
CAS No.: 130723-54-5
Molecular Formula: C8H6IN
Formula Weight: 243.05
Flash Point:
Vzorec:
C₈H₆IN MW: 243,05 g/mol Bod varu: 138 °C (12 mmHg) Bod tání: 33…35 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040890 CAS číslo: 130723-54-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...