Tisk…

4-(Trifluoromethyl)benzenesulphonyl chloride ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-(Trifluoromethyl)benzene-1-sulphonyl chloride

A12907.14 A12907.03 A12907.06
A12907.14EA 9440 CZK
A12907.14 A12907.03 A12907.06
4-(Trifluoromethyl)benzenesulphonyl chloride ≥98%
4-(Trifluoromethyl)benzenesulphonyl chloride

Purity: 98% CAS No.: 2991-42-6 Molecular Formula: C7H4ClF3O2S Formula Weight: 244.62 Flash Point: 110°(230°F) Melting Point: 28-34° Boiling Point: 118-120°/18mm Beilstein Reference: 2113604 UN No.: 3261 MDL No.: MFCD00042422

Vzorec: C₇H₄ClF₃O₂S
MW: 244,62 g/mol
Bod varu: 118…120 °C (18 mmHg)
Bod tání: 28…34 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042422
CAS číslo: 2991-42-6
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR