α,α,α-Trifluoro-m-cresol ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-(Trifluoromethyl)phenol, 3-Hydroxybenzotrifluoride
A12784.06
A12784.14
A12784.22
A12784.06EA
735
CZK
A12784.06
A12784.14
A12784.22
α,α,α-Trifluoro-m-cresol ≥98%
α,α,α-Trifluoro-m-cresol
Purity: 98+%
CAS No.: 98-17-9
Molecular Formula: C7H5F3O
Formula Weight: 162.11
Flash Point: 74°(165°F)
Refractive Index: 1.4580
Odour: Phenol-like
Melting Point: -2°
Boiling Point: 177-178°
Beilstein Reference: 2045663
UN No.: 3265
MDL No.: MFCD00002299
Vzorec:
C₇H₅F₃O MW: 162,11 g/mol Bod varu: 177…178 °C Bod tání: –2 °C Hustota: 1,336 Bod vzplanutí: 74 °C (165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002299 CAS číslo: 98-17-9 EINECS: 202-645-5 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...