2-Fluorobenzyl alcohol ≥98%
Dodavatel: Thermo Fisher Scientific
Varování
|
Synonyms:
(2-Fluorophenyl)methanol, 2-Fluorobenzyl Alcohol
A14791.09
A14791.18
A14791.22
A14791.09EA
1160
CZK
A14791.09
A14791.18
A14791.22
2-Fluorobenzyl alcohol ≥98%
2-Fluorobenzyl alcohol
Purity: 98%
CAS No.: 446-51-5
Molecular Formula: C7H7FO
Formula Weight: 126.13
Flash Point: 90°(194°F)
Refractive Index: 1.5140
Odour: Characteristic
Boiling Point: 201°
Beilstein Reference: 2079486
MDL No.: MFCD00004601
Vzorec:
C₇H₇FO MW: 126.13 Bod varu: 201 °C Hustota: 1.17 Bod vzplanutí: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004601 CAS číslo: 446-51-5 EINECS: 207-170-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...