2,3,4,5,6-Pentafluorobenzyl alcohol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(Perfluorophenyl)methanol, (2,3,4,5,6-Pentafluorophenyl)methanol
A12119.06
A12119.14
A12119.06EA
1500
CZK
A12119.06
A12119.14
2,3,4,5,6-Pentafluorobenzyl alcohol ≥98%
2,3,4,5,6-Pentafluorobenzyl alcohol
Purity: 98%
CAS No.: 440-60-8
Molecular Formula: C7H3F5O
Formula Weight: 198.09
Flash Point: 87°(188°F)
Melting Point: 32-36°
Boiling Point: 182°
Beilstein Reference: 2052669
MDL No.: MFCD00004602
Vzorec:
C₇H₃F₅O MW: 198.09 Bod varu: 182 °C Bod tání: 32…36 °C Bod vzplanutí: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004602 CAS číslo: 440-60-8 EINECS: 207-126-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...