4-Fluoro-m-toluidine ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-Fluoro-3-methylaniline, 5-Amino-2-fluorotoluene
A15482.06
A15482.14
A15482.06EA
1850
CZK
A15482.06
A15482.14
4-Fluoro-m-toluidine ≥97%
4-Fluoro-m-toluidine
Purity: 97%
CAS No.: 452-69-7
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 95°(203°F)
Refractive Index: 1.5370
Odour: Characteristic
Melting Point: 32-36°
Boiling Point: 85-86°/9mm
Beilstein Reference: 2935561
UN No.: 2811
MDL No.: MFCD00025294
Vzorec:
C₇H₈FN MW: 125.15 Bod varu: 85…86 °C (9 mmHg) Bod tání: 32…36 °C Hustota: 1.120 Bod vzplanutí: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025294 CAS číslo: 452-69-7 EINECS: 207-207-7 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...