Tisk…

3-Fluoro-4-(trifluoromethyl)aniline ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Fluoro-4-trifluoromethylaniline, 4-Amino-alpha,alpha,alpha,2-tetrafluorotoluene, α,α,α,3-Tetrafluoro-p-toluidine, 4-Amino-2-fluorobenzotrifluoride, 4-Amino-α,α,α,2-tetrafluorotoluene

B24411.03 B24411.06
B24411.03EA 1040 CZK
B24411.03 B24411.06
3-Fluoro-4-(trifluoromethyl)aniline ≥97%
3-Fluoro-4-(trifluoromethyl)aniline

Purity: 97% CAS No.: 69411-68-3 Molecular Formula: C7H5F4N Formula Weight: 179.12 Melting Point: 54-58° UN No.: 2811 MDL No.: MFCD00190120

Vzorec: C₇H₅F₄N
Bod tání: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00190120
CAS číslo: 69411-68-3
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR