3-Fluoro-o-toluidine ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Amino-6-fluorotoluene, 3-Fluoro-2-methylaniline
B22764.06
B22764.14
B22764.06EA
893
CZK
B22764.06
B22764.14
3-Fluoro-o-toluidine ≥98%
3-Fluoro-o-toluidine
Purity: 98+%
CAS No.: 443-86-7
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 86°(186°F)
Refractive Index: 1.5420
Odour: Characteristic
Melting Point: 7°
Boiling Point: 205°
UN No.: 2810
MDL No.: MFCD00007760
Vzorec:
C₇H₈FN MW: 125,15 g/mol Bod varu: 205 °C Bod tání: 7 °C Hustota: 1,099 Bod vzplanutí: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007760 CAS číslo: 443-86-7 EINECS: 207-142-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...