Tisk…

α,α,α-Trifluoro-m-toluidine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-(Trifluoromethyl)aniline, 3-Aminobenzotrifluoride

M-Clarity™ quality level = MQ100

A41801-100G A41801-500G A41801-5G
SIALA41801-100GEA 1250 CZK
SIALA41801-100G SIALA41801-500G SIALA41801-5G
α,α,α-Trifluoro-m-toluidine, Sigma-Aldrich®
α,α,α-Trifluoro-m-toluidine
Vzorec: C₇H₆F₃N
MW: 161.13
Bod varu: 187
Bod tání: 5
Hustota: 1.3 g/cm³ (20 °C)
Bod vzplanutí: 87
Storage Temperature: Ambient
MDL Number: MFCD00007797
CAS číslo: 98-16-8
EINECS: 202-643-4
UN: 2948
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR