α,α,α-Trifluoro-m-toluidine ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
3-(Trifluoromethyl)aniline, 3-Aminobenzotrifluoride
A15910.0E
A15910.22
A15910.36
A15910.0EEA
10370
CZK
A15910.0E
A15910.22
A15910.36
α,α,α-Trifluoro-m-toluidine ≥99%
α,α,α-Trifluoro-m-toluidine
Purity: 99%
CAS No.: 98-16-8
Molecular Formula: C7H6F3N
Formula Weight: 161.13
Flash Point: 85°(185°F)
Refractive Index: 1.4800
Odour: Recognizable
Melting Point: 5-6°
Boiling Point: 187-189°
Beilstein Reference: 387672
UN No.: 2948
MDL No.: MFCD00007797
Vzorec:
C₇H₆F₃N MW: 161.13 Bod varu: 187…189 °C Bod tání: 5…6 °C Hustota: 1.295 Bod vzplanutí: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007797 CAS číslo: 98-16-8 EINECS: 202-643-4 UN: 2948 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...