Tisk…

6-Fluoro-m-toluidine ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L13411.06 L13411.14
L13411.06EA 769 CZK
L13411.06 L13411.14
6-Fluoro-m-toluidine ≥98%
6-Fluoro-m-toluidine

Purity: 98% CAS No.: 452-84-6 Molecular Formula: C7H8FN Formula Weight: 125.15 Flash Point: 81°(177°F) Refractive Index: 1.5330 Odour: Characteristic Boiling Point: 80-86°/11mm Beilstein Reference: 2715995 UN No.: 2810 MDL No.: MFCD00007654

Vzorec: C₇H₈FN
Bod varu: 80…86 °C (11 mmHg)
Hustota: 1,109
Bod vzplanutí: 81 °C (177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007654
CAS číslo: 452-84-6
UN: 2810
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR