Tisk…

2-Fluoro-p-toluidine ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A16591.06 A16591.14
A16591.06EA 1800 CZK
A16591.06 A16591.14
2-Fluoro-p-toluidine ≥99%
2-Fluoro-p-toluidine

Purity: 99% CAS No.: 452-80-2 Molecular Formula: C7H8FN Formula Weight: 125.15 Flash Point: 79°(174°F) Refractive Index: 1.5330 Boiling Point: 70-71°/7mm Beilstein Reference: 2637578 UN No.: 2810 MDL No.: MFCD00040975

Vzorec: C₇H₈FN
Bod varu: 70…71 °C (7 mmHg)
Bod tání: 3 °C
Hustota: 1,108
Bod vzplanutí: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040975
CAS číslo: 452-80-2
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR