2,6-Difluorobenzylamine, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
264393-1G
264393-5G
SIAL264393-1GEA
1340
CZK
SIAL264393-1G
SIAL264393-5G
2,6-Difluorobenzylamine, Sigma-Aldrich®
2,6-Difluorobenzylamine
Vzorec:
C₇H₇F₂N MW: 143,14 g/mol Bod varu: 174…177 °C (1013 hPa) Hustota: 1,19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010144 CAS číslo: 69385-30-4 EINECS: 273-983-9 UN: 2735 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...