Tisk…
4-(Trifluoromethyl)phenyl isothiocyanate ≥98%
Dodavatel: Thermo Scientific
Nebezpečí
Synonyms: 1-Isocyanato-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-tolyl isothiocyanate, p-Trifluoromethylphenylisothiocyanate
L11890.03 L11890.06
L11890.03EA 1340 CZK
L11890.03 L11890.06
4-(Trifluoromethyl)phenyl isothiocyanate ≥98%
4-(Trifluoromethyl)phenyl isothiocyanate
Purity: 98+% CAS No.: 1645-65-4 Molecular Formula: C8H4F3NS Formula Weight: 203.19 Flash Point: 110°(230°F) Odour: Pungent Boiling Point: 207° Beilstein Reference: 2693553 UN No.: 2923 MDL No.: MFCD00039645
Vzorec: C₈H₄F₃NS
MW: 203,19 g/mol
Bod varu: 207 °C
Bod tání: 39…42 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039645
CAS číslo: 1645-65-4
UN: 2923
ADR: 8,III