α,α,α-Trifluoro-m-tolyl isocyanate ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-(Trifluoromethyl)phenyl isocyanate
L09448.06
L09448.14
L09448.03
L09448.06EA
1780
CZK
L09448.06
L09448.14
L09448.03
α,α,α-Trifluoro-m-tolyl isocyanate ≥98%
α,α,α-Trifluoro-m-tolyl isocyanate
Purity: 98%
CAS No.: 329-01-1
Molecular Formula: C8H4F3NO
Formula Weight: 187.12
Storage Temperature: 2°C to 8°C
Flash Point: 58°(136°F)
Refractive Index: 1.4685
Odour: Pungent
Boiling Point: 53-54°/11mm
Beilstein Reference: 744880
UN No.: 2285
MDL No.: MFCD00002020
Vzorec:
C₈H₄F₃NO MW: 187,12 g/mol Bod varu: 53…54 °C (11 mmHg) Bod tání: –26 °C Hustota: 1,336 Bod vzplanutí: 58 °C (136 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002020 CAS číslo: 329-01-1 EINECS: 206-341-3 UN: 2285 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...