α,α,α-Trifluoro-o-tolyl isocyanate ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-(Trifluoromethyl)phenyl isocyanate, 2-(Trifluoromethyl)phenylisocyanate
L11156.06
L11156.14
L11156.03
L11156.06EA
2060
CZK
L11156.06
L11156.14
L11156.03
α,α,α-Trifluoro-o-tolyl isocyanate ≥97%
α,α,α-Trifluoro-o-tolyl isocyanate
Purity: 97%
CAS No.: 2285-12-3
Molecular Formula: C8H4F3NO
Formula Weight: 187.12
Storage Temperature: 2°C to 8°C
Flash Point: 58°(136°F)
Refractive Index: 1.4755
Odour: Pungent
Boiling Point: 74-75°/40mm
Beilstein Reference: 977056
UN No.: 2285
MDL No.: MFCD00002008
Vzorec:
C₈H₄F₃NO MW: 187,12 g/mol Bod varu: 74…75 °C (40 mmHg) Hustota: 1,238 Bod vzplanutí: 58 °C (136 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002008 CAS číslo: 2285-12-3 EINECS: 218-925-5 UN: 2285 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...