2-Nitro-α,α,α-trifluoro-p-toluic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Nitro-4-(trifluoromethyl)benzoic acid, 2-Nitro-4-trifluoromethylbenzoic acid
43787.09
43787.04
43787.09EA
5080
CZK
43787.09
43787.04
2-Nitro-α,α,α-trifluoro-p-toluic acid ≥98%
2-Nitro-α,α,α-trifluoro-p-toluic acid
Purity: 98%
CAS No.: 320-94-5
Molecular Formula: C8H4F3NO4
Formula Weight: 235.12
Physical Form: Powder
Vzorec:
C₈H₄F₃NO₄ MW: 235,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007142 CAS číslo: 320-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...