Tisk…

5-Amino-α,α,α-trifluoro-m-toluic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

H26169.03 H26169.06
H26169.03EA 2360 CZK
H26169.03 H26169.06
5-Amino-α,α,α-trifluoro-m-toluic acid 97%
5-Amino-α,α,α-trifluoro-m-toluic acid

Purity: 97% CAS No.: 328-68-7 Molecular Formula: C8H6F3NO2 Formula Weight: 205.14 Melting Point: 142-146° MDL No.: MFCD00236641

Vzorec: C₈H₆F₃NO₂
Bod tání: 142…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00236641
CAS číslo: 328-68-7
EINECS: None

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR