Tisk…

4-Fluoro-o-toluic acid, Sigma-Aldrich®

Dodavatel: Merck

638323-1G 638323-5G
SIAL638323-1GEA 0 CZK
SIAL638323-1G SIAL638323-5G
4-Fluoro-o-toluic acid, Sigma-Aldrich®
4-Fluoro-o-toluic acid
Vzorec: C₈H₇FO₂
MW: 154,14 g/mol
MDL Number: MFCD04115880
CAS číslo: 321-21-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR