α,α,α-Trifluoro-p-toluic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-(Trifluoromethyl)benzoic acid
A14494.06
A14494.14
A14494.06EA
1860
CZK
A14494.06
A14494.14
α,α,α-Trifluoro-p-toluic acid ≥98%
α,α,α-Trifluoro-p-toluic acid
Purity: 98+%
CAS No.: 455-24-3
Molecular Formula: C8H5F3O2
Formula Weight: 190.12
Melting Point: 220-222°
Boiling Point: 247°/753mm
MDL No.: MFCD00002562
Vzorec:
C₈H₅F₃O₂ Bod varu: 247 °C (753 mmHg) Bod tání: 220…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002562 CAS číslo: 455-24-3 EINECS: 207-242-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...