3'-Fluoroacetophenone ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-(3-Fluorophenyl)ethanone, m-Fluoroacetophenone
A11733.06
A11733.14
A11733.22
A11733.06EA
1960
CZK
A11733.06
A11733.14
A11733.22
3'-Fluoroacetophenone ≥98%
3'-Fluoroacetophenone
Purity: 98+%
CAS No.: 455-36-7
Molecular Formula: C8H7FO
Formula Weight: 138.14
Flash Point: 80°(176°F)
Refractive Index: 1.5090
Melting Point: -3°
Boiling Point: 80-81°/9mm
Beilstein Reference: 907041
MDL No.: MFCD00000338
Vzorec:
C₈H₇FO MW: 138,14 g/mol Bod varu: 80…81 °C (9 mmHg) Bod tání: –3 °C Hustota: 1,130 Bod vzplanutí: 80 °C (176 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000338 CAS číslo: 455-36-7 EINECS: 207-245-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...