2',4',5'-Trifluoroacetophenone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-(2,4,5-Trifluorophenyl)ethan-1-one, 2,4,5-Trifluoroacetophenone
M-Clarity™ quality level = MQ100
340804-1G
340804-5G
SIAL340804-1GEA
1820
CZK
SIAL340804-1G
SIAL340804-5G
2',4',5'-Trifluoroacetophenone, Sigma-Aldrich®
2',4',5'-Trifluoroacetophenone
Vzorec:
C₈H₅F₃O MW: 174,12 g/mol Bod varu: 76 °C (1013 hPa) Hustota: 1,38 g/cm³ (20 °C) |
MDL Number:
MFCD00061193 CAS číslo: 129322-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...