Tisk…

3',5'-Difluoroacetophenone ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-(3,5-Difluorophenyl)ethan-1-one

B20301.03 B20301.14
B20301.03EA 742 CZK
B20301.03 B20301.14
3',5'-Difluoroacetophenone ≥97%
3',5'-Difluoroacetophenone

Purity: 97% CAS No.: 123577-99-1 Molecular Formula: C8H6F2O Formula Weight: 156.13 Flash Point: 82°(179°F) Refractive Index: 1.4876 Melting Point: 37-41° Boiling Point: 94-95°/13mm Beilstein Reference: 6798740 MDL No.: MFCD00042489

Vzorec: C₈H₆F₂O
MW: 156,13 g/mol
Bod varu: 94…95 °C (13 mmHg)
Bod tání: 37…41 °C
Bod vzplanutí: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042489
CAS číslo: 123577-99-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR