Tisk…

2',4'-Difluoroacetophenone ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-(2,4-Difluorophenyl)ethan-1-one

A13420.06 A13420.14
A13420.06EA 1780 CZK
A13420.06 A13420.14
2',4'-Difluoroacetophenone ≥98%
2',4'-Difluoroacetophenone

Purity: 98% CAS No.: 364-83-0 Molecular Formula: C8H6F2O Formula Weight: 156.13 Flash Point: 66°(150°F) Refractive Index: 1.4880 Odour: Characteristic Boiling Point: 80-81°/25mm Beilstein Reference: 1940741 MDL No.: MFCD00151261

Vzorec: C₈H₆F₂O
MW: 156,13 g/mol
Bod varu: 80…81 °C (25 mmHg)
Hustota: 1,232
Bod vzplanutí: 66 °C (150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00151261
CAS číslo: 364-83-0
EINECS: 206-667-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR