Tisk…

2',6'-Difluoroacetophenone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-(2,6-Difluorophenyl)ethan-1-one, 1-(2,6-Difluorophenyl)ethanone

M-Clarity™ quality level = MQ100

232890-1G 232890-5G
SIAL232890-1GEA 1630 CZK
SIAL232890-1G SIAL232890-5G
2',6'-Difluoroacetophenone, Sigma-Aldrich®
2',6'-Difluoroacetophenone
Vzorec: C₈H₆F₂O
MW: 156,13 g/mol
Bod varu: 76…79 °C (15 mmHg)
Hustota: 1,19 g/cm³ (20 °C)
MDL Number: MFCD00000328
CAS číslo: 13670-99-0
EINECS: 237-151-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR