2',3'-Difluoroacetophenone ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1-(2,3-Difluorophenyl)ethan-1-one, 1-(2,3-Difluorophenyl)ethanone, 2,3-Difluoroacetophenone
A16786.06
A16786.14
A16786.06EA
2650
CZK
A16786.06
A16786.14
2',3'-Difluoroacetophenone ≥97%
2',3'-Difluoroacetophenone
Purity: 97%
CAS No.: 18355-80-1
Molecular Formula: C8H6F2O
Formula Weight: 156.13
Refractive Index: 1.4890
Odour: Characteristic
Boiling Point: 84-85°/20mm
Beilstein Reference: 2963572
MDL No.: MFCD00061274
Vzorec:
C₈H₆F₂O MW: 156.13 Bod varu: 84…85 °C (20 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00061274 CAS číslo: 18355-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...