(4-Fluorophenyl)acetonitrile, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Fluorobenzyl cyanide, 4-Fluorophenylacetonitrile
M-Clarity™ quality level = MQ100
102407-100G
102407-25G
SIAL102407-100GEA
6230
CZK
SIAL102407-100G
SIAL102407-25G
(4-Fluorophenyl)acetonitrile, Sigma-Aldrich®
(4-Fluorophenyl)acetonitrile
Vzorec:
C₈H₆FN MW: 135,14 g/mol Bod varu: 234 °C (1013 hPa) Bod tání: 86 °C Hustota: 1,136 g/cm³ (20 °C) |
MDL Number:
MFCD00001917 CAS číslo: 459-22-3 EINECS: 207-286-8 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...