(2-Fluorophenyl)acetonitrile ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Fluorobenzyl cyanide, 2-(2-Fluorophenyl)acetonitrile
A11612.06
A11612.14
A11612.22
A11612.06EA
865
CZK
A11612.06
A11612.14
A11612.22
(2-Fluorophenyl)acetonitrile ≥97%
(2-Fluorophenyl)acetonitrile
Purity: 97%
CAS No.: 326-62-5
Molecular Formula: C8H6FN
Formula Weight: 135.14
Flash Point:
Vzorec:
C₈H₆FN MW: 135,14 g/mol Bod varu: 235 °C Hustota: 1,061 Bod vzplanutí: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001897 CAS číslo: 326-62-5 EINECS: 206-313-0 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...