4-(Trifluoromethyl)phenylboronic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-(Trifluoromethyl)benzeneboronic acid, (4-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-(trifluoromethyl)phenyl)borane, (4-(Trifluoromethyl)phenyl)boronic acid
B22374.03
B22374.06
B22374.14
B22374.03EA
1600
CZK
B22374.03
B22374.06
B22374.14
4-(Trifluoromethyl)phenylboronic acid ≥98%
4-(Trifluoromethyl)phenylboronic acid
Purity: 98%
CAS No.: 128796-39-4
Molecular Formula: C7H6BF3O2
Formula Weight: 189.93
Melting Point: 238-242°
Beilstein Reference: 3544189
Vzorec:
C₇H₆BF₃O₂ MW: 189,93 g/mol Bod tání: 238…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151855 CAS číslo: 128796-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...