3,4-Difluorotoluene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
41789.03
41789.06
41789.03EA
1210
CZK
41789.03
41789.06
3,4-Difluorotoluene ≥98%
3,4-Difluorotoluene
Purity: 98%
CAS No.: 2927-34-6
Molecular Formula: C7H6F2
Formula Weight: 128.12
Flash Point: 25°(77°F)
Physical Form: Liquid
Density: 1.120
Refractive Index: 1.4500
Odour: Aromatic
Boiling Point: 110-113°
UN No.: 1993
MDL No.: MFCD00075087
Vzorec:
C₇H₆F₂ Bod varu: 110…113 °C Hustota: 1,120 Bod vzplanutí: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075087 CAS číslo: 2927-34-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...