Tisk…

2-Hydroxy-p-toluic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 2-Hydroxy-4-methylbenzoic acid, m-Homosalicyclic acid, m-Cresotic acid, 4-Methylsalicylic acid

M-Clarity™ quality level = MQ100

244503-100G 244503-25G
SIAL244503-100GEA 4970 CZK
SIAL244503-100G SIAL244503-25G
2-Hydroxy-p-toluic acid, Sigma-Aldrich®
2-Hydroxy-p-toluic acid
Vzorec: C₈H₈O₃
MW: 152.15
Bod tání: 177
MDL Number: MFCD00002454
CAS číslo: 50-85-1
EINECS: 200-068-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR