4-Methyl-m-anisic acid ≥99%
Dodavatel: Thermo Fisher Scientific
B22982.06
B22982.14
B22982.06EA
2670
CZK
B22982.06
B22982.14
4-Methyl-m-anisic acid ≥99%
4-Methyl-m-anisic acid
Purity: 99%
CAS No.: 7151-68-0
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 158-160°
Beilstein Reference: 1938389
Vzorec:
C₉H₁₀O₃ Bod tání: 158…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002566 CAS číslo: 7151-68-0 EINECS: 230-486-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...