Tisk…

4-Ethylbenzoic acid, Sigma-Aldrich®

Dodavatel: Merck

191280-25G 191280-5G
SIAL191280-25GEA 3590 CZK
SIAL191280-25G SIAL191280-5G
4-Ethylbenzoic acid, Sigma-Aldrich®
4-Ethylbenzoic acid
Vzorec: C₉H₁₀O₂
MW: 150,18 g/mol
MDL Number: MFCD00002570
CAS číslo: 619-64-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR