4-Hexylbenzoyl chloride ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A15659.06
A15659.14
A15659.22
A15659.06EA
2160
CZK
A15659.06
A15659.14
A15659.22
4-Hexylbenzoyl chloride ≥98%
4-Hexylbenzoyl chloride
Purity: 98%
CAS No.: 50606-95-6
Molecular Formula: C13H17ClO
Formula Weight: 224.73
Flash Point:
Vzorec:
C₁₃H₁₇ClO Bod varu: 176…177 °C (22 mmHg) Hustota: 1,029 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000701 CAS číslo: 50606-95-6 EINECS: 256-647-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...