4-Ethylbenzoyl chloride ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-Ethylbenzene-1-carbonyl chloride, 4-Ethylbenzoylchloride
A11698.06
A11698.09
A11698.18
A11698.06EA
917
CZK
A11698.06
A11698.09
A11698.18
4-Ethylbenzoyl chloride ≥98%
4-Ethylbenzoyl chloride
Purity: 98+%
CAS No.: 16331-45-6
Molecular Formula: C9H9ClO
Formula Weight: 168.62
Flash Point:
Vzorec:
C₉H₉ClO Bod varu: 235…236 °C Bod tání: 42…43 °C Hustota: 1,146 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000697 CAS číslo: 16331-45-6 EINECS: 240-404-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...