Tisk…

4-Decylbenzoyl chloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A17234.06 A17234.14
A17234.06EA 5430 CZK
A17234.06 A17234.14
4-Decylbenzoyl chloride ≥98%
4-Decylbenzoyl chloride

Purity: 98% CAS No.: 54256-43-8 Molecular Formula: C17H25ClO Formula Weight: 280.84 Flash Point: 110°(230°F) Boiling Point: 171-173°/2mm UN No.: 3265 MDL No.: MFCD00041724

Vzorec: C₁₇H₂₅ClO
Bod varu: 171…173 °C (2 mmHg)
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041724
CAS číslo: 54256-43-8
EINECS: 259-046-7
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR