3-Toluoyl chloride ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-Methylbenzoyl chloride, m-Methylbenzoyl chloride, m-Toluoyl chloride, 3-Methylbenzene-1-carbonyl chloride
B23914.14
B23914.22
B23914.36
B23914.14EA
1080
CZK
B23914.14
B23914.22
B23914.36
3-Toluoyl chloride ≥99%
3-Toluoyl chloride
Purity: 99%
CAS No.: 1711-06-4
Molecular Formula: C8H7ClO
Formula Weight: 154.60
Flash Point: 76°(168°F)
Refractive Index: 1.5485
Odour: Pungent
Melting Point: -23°
Boiling Point: 220°
Beilstein Reference: 878419
UN No.: 3265
MDL No.: MFCD00000681
Vzorec:
C₈H₇ClO MW: 154,6 g/mol Bod varu: 220 °C Bod tání: –23 °C Hustota: 1,173 Bod vzplanutí: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000681 CAS číslo: 1711-06-4 EINECS: 216-976-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...