Tisk…

3-Toluoyl chloride 99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3-Methylbenzoyl chloride, m-Methylbenzoyl chloride, m-Toluoyl chloride, 3-Methylbenzene-1-carbonyl chloride

153410050 153411000 153415000
ACRO153410050EA 1070 CZK
ACRO153410050 ACRO153411000 ACRO153415000
3-Toluoyl chloride 99%
3-Toluoyl chloride
Vzorec: C₈H₇ClO
MW: 154,6 g/mol
Bod varu: 86 °C (5 mmHg)
Bod tání: –23 °C
Hustota: 1,17 g/cm³
Bod vzplanutí: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00000681
CAS číslo: 1711-06-4
EINECS: 216-976-8
UN: 3265
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5475 to 1.5495 (20°C, 589 nm)
Titration RCOCl ≥98.5 %
UV at 280 nm A: ≤0.050
pH 10.9 to 11.6 (1 % in water at 25°C)
Titration with HClO4 97.5 to 102.5 % (on dry basis)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR