3',5'-Dihydroxyacetophenone 96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-(3,5-Dihydroxyphenyl)ethan-1-one, 1-(3,5-Dihydroxyphenyl)ethanone
B25567.09
B25567.18
B25567.09EA
3880
CZK
B25567.09
B25567.18
3',5'-Dihydroxyacetophenone 96%
3',5'-Dihydroxyacetophenone
Purity: 96%
CAS No.: 51863-60-6
Molecular Formula: C8H8O3
Formula Weight: 152.15
Melting Point: 142-149°
Beilstein Reference: 1936502
Vzorec:
C₈H₈O₃ MW: 152,15 g/mol Bod varu: 296 °C (1013 hPa) Bod tání: 142…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002290 CAS číslo: 51863-60-6 EINECS: 257-480-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...