2',6'-Dihydroxyacetophenone ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-(2,6-Dihydroxyphenyl)ethan-1-one, 1-(2,6-Dihydroxyphenyl)ethanone, 2,6-Dihydroxyacetophenone
A14627.06
A14627.14
A14627.22
A14627.06EA
1090
CZK
A14627.06
A14627.14
A14627.22
2',6'-Dihydroxyacetophenone ≥98%
2',6'-Dihydroxyacetophenone
Purity: 98+%
CAS No.: 699-83-2
Molecular Formula: C8H8O3
Formula Weight: 152.15
Melting Point: 156-159°
Beilstein Reference: 1366061
Vzorec:
C₈H₈O₃ MW: 152,15 g/mol Bod varu: 296 °C (1013 hPa) Bod tání: 156…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002270 CAS číslo: 699-83-2 EINECS: 211-833-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...